Orders LinkContactUsLinkComponent
Adapters, expansion
Adapters, expansion
Expansion, 14/23 NS, 29/32 NS
Catalog # WITG0.510.003
Supplier:   witeg Labortechnik
Adapters, expansion
Catalog # WITG0.510.003
Supplier:  witeg Labortechnik
Adapters, expansion
Adapters, expansion
Expansion, 14/23 NS, 29/32 NS
Price
14,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapters.
Certifications: According to DIN 12257.
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    29/32 NS
  • Type:
    Expansion
  • Seq:
    02