5-Nitroindole
Glass bottle for solids, 50 g
MAYBSB01605.50
Marketsource
: 5-Nitro-1H-indole
- :6146-52-7
- :C₈H₆N₂O₂
- :MFCD00005673
- :162.15 g/mol
- :140 °C
- :228-153-0
- Pack type:Glass bottle for solids
- Pk:50 g