Order Entry
Belgium
Orders LinkContactUsLinkComponent
Water

Water

Glass bottle, 500 mL
Catalog # AMBI9920
Marketsource
Supplier:   Ambion
Water
Catalog # AMBI9920
Supplier:  Ambion
Marketsource
CAS Number:  7732-18-5
Water

Glass bottle, 500 mL

CAS Number:  7732-18-5
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications