Orders LinkContactUsLinkComponent
Multiple or single cuvette segments
Multiple or single cuvette segments
Cuvettes in bulk segments, PMMA
Catalog # KART1501
Supplier:   KARTELL
Multiple or single cuvette segments
Catalog # KART1501
Supplier:  KARTELL
Multiple or single cuvette segments
Multiple or single cuvette segments
Cuvettes in bulk segments, PMMA
Price
794,00 €
Log in for availability
1 * 450 ST
Product Details & Documents
These twelve position cuvettes segments have been specially designed for use with the Cobas Mira® analyser.
Packaging: In cases of bulk segments (450) or 30 pre-racked segments (15).
Specifications
  • Description:
    Cuvettes in bulk segments
  • Material:
    PMMA