Orders LinkContactUsLinkComponent
HPLC columns, ZORBAX® Eclipse XDB
HPLC columns, ZORBAX® Eclipse XDB
Rapid resolution HT cartridge, Guard cartridge, Eclipse XDB-C18, 1,8 µm, 2,10 mm, 15 mm
  HEWL921700-932
 :   Agilent
HPLC columns, ZORBAX® Eclipse XDB
  HEWL921700-932
 :  Agilent
HPLC columns, ZORBAX® Eclipse XDB
HPLC columns, ZORBAX® Eclipse XDB
Rapid resolution HT cartridge, Guard cartridge, Eclipse XDB-C18, 1,8 µm, 2,10 mm, 15 mm
1 010,00 €
1 * 3 ST
Good peak shape for basic, acidic and neutral compounds.
  • High performance over a wide pH range (2 to 9)
  • Long lifetime with extra dens bonding and double endcapping
  • Column Format:
    Guard cartridge
  • Ø int.:
    2,10 mm
  • Particle Size:
    1,8 µm
  • Description:
    Rapid resolution HT cartridge
  • Length:
    15 mm
  • Phase:
    Eclipse XDB-C18