Orders LinkContactUsLinkComponent
Solvent filter/degasser
Solvent filter/degasser
PTFE seal for HEWL3150-0577, PTFE
Catalog # HEWL5188-2745
Supplier:   Agilent
Solvent filter/degasser
Catalog # HEWL5188-2745
Supplier:  Agilent
Solvent filter/degasser
Solvent filter/degasser
PTFE seal for HEWL3150-0577, PTFE
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
  • Degasses eluents as particulates are removed
  • Increases solvent inlet lifetime
  • Eliminates pump downtime caused by air locks and particulates in check valves
  • Decreases piston wear, while increasing column life
Specifications
  • Description:
    PTFE seal for HEWL3150-0577
  • Material:
    PTFE
  • Seq:
    05