CD4 MICROBEADS HUMAN
Catalog # MILT130-045-101
Supplier: Miltenyi Biotec
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™ Xtra™
Supplier: KIMBERLY CLARK
Ethyl acetate ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Xylene (mixture of isomers) ≥98%, TECHNICAL
Supplier: VWR Chemicals
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Xylene (mixture of isomers) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit
Supplier: STEMCELL Technologies
Tetrahydrofuran, extra dry over Molecular Sieve 99.5% stabilised, AcroSeal®
Supplier: Thermo Fisher Scientific
U-PLEX Proinflam Combo 2 (human) 1 * 5 items
Supplier: MESO SCALE DISCOVERY