PRE-SEPARATION FILTERS
Catalog # MILT130-041-407
Supplier: Miltenyi Biotec
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Accessories for ThermoMixer™ F1,5 and FP
Supplier: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Spray bottles, LaboPlast®
Supplier: BURKLE
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Cryoboxes and Dividers
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
U-PLEX PROINFLAM COMBO 1 (HU) SECTOR 1 * 1 items
Supplier: MESO SCALE DISCOVERY
Seal and aluminium foil lids, Biomek®
Supplier: Beckman Coulter
[EN]BLITZ 10X KINETIC BUFFER PROCLIN 1X
Supplier: PALL FORTEBIO
REAGENT, SPRISELECT, 60 ML 1 * 60 mL
Supplier: Beckman Coulter