Order Entry
Belgium
Orders LinkContactUsLinkComponent
RevertAid™ Reverse Transcriptase

RevertAid™ Reverse Transcriptase

RevertAid™ Reverse Transcriptase, 5×10,000 units (200 U/µl), for 250 reactions of 20 µl, 1 KIT
Catalog # EP0442
RevertAid™ Reverse Transcriptase
Catalog # EP0442
CAS Number:  
RevertAid™ Reverse Transcriptase
RevertAid™ Reverse Transcriptase

RevertAid™ Reverse Transcriptase, 5×10,000 units (200 U/µl), for 250 reactions of 20 µl, 1 KIT

CAS Number:  
Price
373,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

RevertAid™ Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids, which is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.

  • Efficient synthesis of full length first strand cDNA up to 13 kb
  • Optimum activity at 42 °C
  • Active up to 50 °C
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labelled nucleotides)

Delivery information: Supplied with 5X reaction buffer.

Documents

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    RevertAid™ Reverse Transcriptase, 5×10,000 units (200 U/µl), for 250 reactions of 20 µl

Specifications