Order Entry
Belgium
Orders LinkContactUsLinkComponent
RevertAid™ Reverse Transcriptase

RevertAid™ Reverse Transcriptase

RevertAid™ Reverse Transcriptase, 10,000 units (200 U/µl), for 50 reactions of 20 µl, 1 KIT
Catalog # EP0441
RevertAid™ Reverse Transcriptase
Catalog # EP0441
CAS Number:  
RevertAid™ Reverse Transcriptase
RevertAid™ Reverse Transcriptase

RevertAid™ Reverse Transcriptase, 10,000 units (200 U/µl), for 50 reactions of 20 µl, 1 KIT

CAS Number:  
Price
98,30 €
Log in for availability
1 * 1 KIT
Product Details & Documents

RevertAid™ Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids, which is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.

  • Efficient synthesis of full length first strand cDNA up to 13 kb
  • Optimum activity at 42 °C
  • Active up to 50 °C
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labelled nucleotides)

Delivery information: Supplied with 5X reaction buffer.

Documents

Specifications

Specifications

  • Pk:
    1 KIT
  • Description:
    RevertAid™ Reverse Transcriptase, 10,000 units (200 U/µl), for 50 reactions of 20 µl

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.