RevertAid™ H Minus Reverse Transcriptase
Product Details & Documents
RevertAid™ H Minus Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity, but lacks RNase H activity due to point mutation in the RNase H domain. RevertAid™ H Minus Reverse Transcriptase does not degrade RNA in RNA-DNA hybrids during synthesis of the first strand cDNA and therefore high yields of full-length cDNA from long templates are obtained. RevertAid™ H Minus Reverse Transcriptase maintains activity over a wide temperature range (42 to 55 °C), which makes the enzyme an ideal tool for reverse transcription of RNAs having a high degree of secondary structure.
- High yields of full length first strand cDNA up to 13 kb
- Optimum activity at 42 to 45 °C
- Active up to 55 °C
- Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labelled nucleotides)
Delivery information: Supplied with 5X reaction buffer.
Specifications
Specifications
- Cdt:1 KIT
- Désignation:RevertAid™ H Minus Reverse Transcriptase, 5×10 000 units (200 U/µl), for 250 reactions of 20 µl
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



