Order Entry
Belgium
Orders LinkContactUsLinkComponent
Digitonin

Digitonin

Glass bottle for solids, 500 mg
Catalog # SERA19550.01
Marketsource
Supplier:   Serva
Digitonin
Catalog # SERA19550.01
Supplier:  Serva
Marketsource
CAS Number:  11024-24-1
Digitonin

Glass bottle for solids, 500 mg

CAS Number:  11024-24-1
Price
Inquire For Price
Log in for availability
1 * 500 mg
Product Details & Documents
Synonyms:  Digitonin
  • CAS Number:11024-24-1
  • Formula:C₅₆H₉₂O₂₉
  • MDL Number:MFCD00077729
  • Molecular Weight:1229.312 g/mol
  • EINECS:234-255-6
  • UN:3462
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.