Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Agarose

Glass bottle for solids, 100 g
Catalog # SERA11380.02
Marketsource
Supplier:   Serva
Agarose
Catalog # SERA11380.02
Supplier:  Serva
Marketsource
CAS Number:  9012-36-6
Agarose
Agarose

Glass bottle for solids, 100 g

CAS Number:  9012-36-6
Price
130,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:9012-36-6
  • MDL Number:MFCD00081294
  • Melting Pt:60…90 °C
  • Storage Temperature:Ambient
  • EINECS:232-731-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications