Orders LinkContactUsLinkComponent
Base materials
Base materials
Peptone from meat (pancreatic digest), Glass bottle for solids, 250 g
Catalog # SERA48619.01
Marketsource
Supplier:   Serva
Base materials
Catalog # SERA48619.01
Supplier:  Serva
Marketsource
CAS Number:  
Base materials
Peptone from meat (pancreatic digest), Glass bottle for solids, 250 g
CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 250 g
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
  • Suitable as nutrient for laboratory media and industrial fermentation
Specifications
Specifications
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g
  • Description:
    Peptone from meat (pancreatic digest)