Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer), 10 L
Catalog # SERA42553.04
Marketsource
Supplier: Serva
Buffer, TAE solution 10X concentrate (TRIS-acetate-EDTA buffer)
Catalog # SERA42553.04
Supplier: Serva
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:10 L
- Description:TAE buffer solution 10X concentrate (TRIS-acetate-EDTA buffer)



