Order Entry
Belgium
Orders LinkContactUsLinkComponent
Single-component explosives solutions

Single-component explosives solutions

Internal standard includes nitroguanidine, with 1000 µg/ml in methanol, 1 ml/ampule, 1 mL
Catalog # REST31602
Supplier:   Restek
Single-component explosives solutions
Catalog # REST31602
Supplier:  Restek
CAS Number:  
Single-component explosives solutions
Single-component explosives solutions

Internal standard includes nitroguanidine, with 1000 µg/ml in methanol, 1 ml/ampule, 1 mL

CAS Number:  
Price
42,80 €
Log in for availability
1 * 1 mL
Product Details & Documents

Single-component explosives solutions supports US department of defense base closures and remediation.

  • Mixtures and singles supporting LC US EPA method 8330
  • Mixtures and singles supporting GC-ECD US EPA method 8095
  • Internal standards and surrogates to support both methods

These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Internal standard includes nitroguanidine, with 1000 µg/ml in methanol, 1 ml/ampule

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.