Single-component explosives solutions
Internal standard includes 1,2-dinitrotoluene, with 1000 µg/ml in methanol, 1 ml/ampule, 1 mL
Catalog # REST31453
Supplier: Restek
Jump to:
Product Details & Documents
Single-component explosives solutions supports US department of defense base closures and remediation.
- Mixtures and singles supporting LC US EPA method 8330
- Mixtures and singles supporting GC-ECD US EPA method 8095
- Internal standards and surrogates to support both methods
These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.
Specifications
Specifications
- Pk:1 mL
- Description:Internal standard includes 1,2-dinitrotoluene, with 1000 µg/ml in methanol, 1 ml/ampule
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



