Aroclor solutions
Aroclor 1242, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule, 1 mL
Catalog # REST32081
Supplier: Restek
Jump to:
Product Details & Documents
Aroclor standards may exhibit lot-to-lot variation.
: CRM
Specifications
Specifications
- Pk:1 mL
- Description:Aroclor 1242, 50 mg/kg, transformer oil (PCB free), 1 ml/ampoule
- Cat. No.:REST32081
- Formula:C₁₂H₆Cl₄
- Description:Aroclor 1242
- CAS No.:53469-21-9



