Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Aroclor solutions

Aroclor 1254, 200 µg/ml, isooctane, 1 ml/ampoule, 1 mL
Catalog # REST32069
Supplier:   Restek
Aroclor solutions
Catalog # REST32069
Supplier:  Restek
CAS Number:  
Aroclor solutions
Aroclor solutions

Aroclor 1254, 200 µg/ml, isooctane, 1 ml/ampoule, 1 mL

CAS Number:  
Price
50,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Aroclor standards may exhibit lot-to-lot variation.

 : CRM

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1254, 200 µg/ml, isooctane, 1 ml/ampoule
  • Cat. No.:
    REST32069
  • Formula:
    C₁₂H₅Cl₅
  • Description:
    Aroclor 1254
  • CAS No.:
    11097-69-1

Specifications