Order Entry
Belgium
Orders LinkContactUsLinkComponent
Aroclor solutions

Aroclor solutions

Aroclor 1221, 200 µg/ml, isooctane, 1 ml/ampoule, 1 mL
Catalog # REST32065
Supplier:   Restek
Aroclor solutions
Catalog # REST32065
Supplier:  Restek
CAS Number:  
Aroclor solutions
Aroclor solutions

Aroclor 1221, 200 µg/ml, isooctane, 1 ml/ampoule, 1 mL

CAS Number:  
Price
50,00 €
Connexion Pour la disponibilité
1 * 1 mL
Product Details & Documents

Aroclor standards may exhibit lot-to-lot variation.

Certifications: CRM

Spécifications

Spécifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1221, 200 µg/ml, isooctane, 1 ml/ampoule
  • Cat. No.:
    REST32065
  • Formula:
  • Description:
    Aroclor 1221
  • CAS No.:
    11104-28-2

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.