Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Aroclor solutions

Aroclor 1016, 200 µg/ml, isooctane, 1 ml/ampoule, 1 mL
Catalog # REST32064
Supplier:   Restek
Aroclor solutions
Catalog # REST32064
Supplier:  Restek
CAS Number:  
Aroclor solutions
Aroclor solutions

Aroclor 1016, 200 µg/ml, isooctane, 1 ml/ampoule, 1 mL

CAS Number:  
Price
50,00 €
Log in for availability
1 * 1 mL
Product Details & Documents

Aroclor standards may exhibit lot-to-lot variation.

Certifications: CRM

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1016, 200 µg/ml, isooctane, 1 ml/ampoule
  • Cat. No.:
    REST32064
  • Formula:
    C₁₂H₇Cl₃
  • Description:
    Aroclor 1016
  • CAS No.:
    12674-11-2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.