Order Entry
Belgium
Orders LinkContactUsLinkComponent
Aroclor solutions

Aroclor solutions

Aroclor 1260, 1000 µg/ml, hexane, 1 ml/ampoule, 1 mL
Catalog # REST32012
Supplier:   Restek
Aroclor solutions
Catalog # REST32012
Supplier:  Restek
CAS Number:  
Aroclor solutions
Aroclor solutions

Aroclor 1260, 1000 µg/ml, hexane, 1 ml/ampoule, 1 mL

CAS Number:  
Price
70,60 €
Log in for availability
1 * 1 mL
Product Details & Documents

Aroclor standards may exhibit lot-to-lot variation.

Certifications: CRM

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1260, 1000 µg/ml, hexane, 1 ml/ampoule
  • Cat. No.:
    REST32012
  • Formula:
    C₁₂H₄Cl₆
  • Description:
    Aroclor 1260
  • CAS No.:
    11096-82-5

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.