Order Entry
Belgium
Orders LinkContactUsLinkComponent
Aroclor solutions

Aroclor solutions

Aroclor 1242, 1000 µg/ml, hexane, 1 ml/ampoule, 1 mL
Catalog # REST32009
Supplier:   Restek
Aroclor solutions
Catalog # REST32009
Supplier:  Restek
CAS Number:  
Aroclor solutions
Aroclor solutions

Aroclor 1242, 1000 µg/ml, hexane, 1 ml/ampoule, 1 mL

CAS Number:  
Price
69,60 €
Log in for availability
1 * 1 mL
Product Details & Documents

Aroclor standards may exhibit lot-to-lot variation.

Certifications: CRM

Specifications

Specifications

  • Pk:
    1 mL
  • Description:
    Aroclor 1242, 1000 µg/ml, hexane, 1 ml/ampoule
  • Cat. No.:
    REST32009
  • Formula:
    C₁₂H₆Cl₄
  • Description:
    Aroclor 1242
  • CAS No.:
    53469-21-9

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.