VWR® Rectangular Cover Glasses
24×40 mm, #1
631-0145
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.
- No.:#1
- L×W:24×40 mm
- Seq:11
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.