γ-Cyclodextrine
Glass bottle for solids, 500 mg
Catalog # ICNA02190054.5
Supplier: MP Biomedicals
Jump to:
Product Details & Documents
Synonyms: gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
- CAS Number:17465-86-0
- Formula:C₄₈H₈₀O₄₀
- MDL Number:MFCD00009595
- Molecular Weight:1297.1248 g/mol
- EINECS:241-482-4
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



