Orders LinkContactUsLinkComponent
pH indicator papers, Supelco®
pH indicator papers, Supelco®
Acilit®, 0,5 - 5,0, 0,5
N° de catalogue 1.09560.0003
Fournisseur:   Merck
pH indicator papers, Supelco®
N° de catalogue 1.09560.0003
Fournisseur:  Merck
pH indicator papers, Supelco®
pH indicator papers, Supelco®
Acilit®, 0,5 - 5,0, 0,5
Prix
57,50 €
Log in for availability
1 * 3 ROU
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
  • Roll format are easy to use and most common ranges are available in a refill format
  • Quality assurance - tests are checked and calibrated against certified buffer solutions
  • Wide and narrow pH ranges available
Specifications
  • pH gradation:
    0,5
  • pH range:
    0,5 - 5,0
  • Description:
    Acilit®
  • Cat. No.:
    1.09560.0003
  • M-Clarity quality level:
    MQ100