Orders LinkContactUsLinkComponent
Hexagonal nuts with inner thread
Hexagonal nuts with inner thread
hexagonal nut with inner thread, for Ø 27 mm, ½", PE
Catalog # BURK8537-0012
Supplier:   BURKLE
Hexagonal nuts with inner thread
Catalog # BURK8537-0012
Supplier:  BURKLE
Hexagonal nuts with inner thread
Hexagonal nuts with inner thread
hexagonal nut with inner thread, for Ø 27 mm, ½", PE
Price
4,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
HDPE, white.
  • Good chemical resistance
  • Good temperature resistance
Hexagonal nut with inner thread.
Specifications
  • Description:
    hexagonal nut with inner thread, for Ø 27 mm
  • Material:
    PE
  • Thread:
    ½"