Order Entry
Belgium
Orders LinkContactUsLinkComponent
Volumetric flasks

Volumetric flasks

50 ml, GL 18, 143 mm, 0,06 ± ml
Catalog # 612-4919
Supplier:   Brand
Volumetric flasks
Catalog # 612-4919
Supplier:  Brand
Volumetric flasks
Volumetric flasks

50 ml, GL 18, 143 mm, 0,06 ± ml

Price
180,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

PFA, translucent, with PFA screw cap.

  • Inclusive one batch certificate
  • Exposure to temperatures up to 121 ºC (autoclaving) will not cause permanently exceeded error limits
  • Cleaning up to max. 60 °C is recommended to preserve marks and inscriptions

Error limits according to class A, DIN EN ISO 1042. Calibrated to contain (TC, In). PFA screw cap with molded sealing ring. Autoclavable, easy to clean.

*Wide neck.

(Height = Height without stopper).

Specifications
  • Thread:
    GL 18
  • Capacity:
    50 ml
  • Height:
    143 mm
  • Tolerance:
    0,06 ± ml