Orders LinkContactUsLinkComponent
Microscope slides, Menzel Gläser, SuperFrost®
Microscope slides, Menzel Gläser, SuperFrost®
Adhesive slide, SuperFrost®, Blue
Catalog # 631-0706
Supplier:   Epredia
Microscope slides, Menzel Gläser, SuperFrost®
Catalog # 631-0706
Supplier:  Epredia
Microscope slides, Menzel Gläser, SuperFrost®
Microscope slides, Menzel Gläser, SuperFrost®
Adhesive slide, SuperFrost®, Blue
Price
8,40 €
Log in for availability
1 * 50 ST
Product Details & Documents
These slides come with a light opaque coating in standard colours.
  • Ground 90° edges
  • Label areas for easy identification
  • Resistant to solvents, and to scratching or sticking when stacked together
Each colour codes different specimens at a glance according to urgency, processing or staining technique.
Documents
Specifications
  • Colour:
    Blue
  • Description:
    Adhesive slide, SuperFrost®