Cell culture chambers, Corning® CellSTACK®
Product Details & Documents
PS, sterile. The Corning® CellSTACK® culture chambers are available in five sizes and with three different surface treatments.
- Two 26 mm diameter filling ports allow direct access to chamber bottom providing greater flexibility for sterile filling and emptying by pouring, pipetting or via tubing in a fully closed system
- Standard 33 mm threaded caps have 0,2 μm pore non-wettable membranes sealed directly to the caps to allow gas exchange while minimising the risk of contamination
- Optional 33 mm threaded caps are available with integrally sealed USP Class VI certified C-Flex® tubing to allow direct sterile transfer of media and cells via pumping or gravity feed
- Polystyrene construction provides excellent optical clarity and mechanical strength
TC treated surface: Uncharged polystyrene has an uncharged, hydrophobic surface to which cell attachment proteins bind poorly. This results in poor and uneven cell attachment and growth. Tissue culture (TC) treated polystyrene has a negatively charged, hydrophilic surface to which cell attachment proteins bind evenly. This provides a good surface for cell attachment and growth.
Corning® CellBIND® surface: The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields.
More consistent cell attachment resulting in increased cell growth
Adapts cells more quickly to reduced-serum or serum-free conditions
Reduces premature cell detachment from confluent cultures
May eliminate the need for tedious, time-consuming, low stability biological coatings
Requires no refrigeration or special handling and is stable at room temperature
Ultra-Low Attachment surface: The Ultra-Low Attachment surface is a covalently bonded hydrogel surface that is hydrophilic and neutrally charged. It minimises cell attachment, protein absorption and enzyme activation. The surface is non-cytotoxic, biologically inert and non-degradable.
Maintains cells in a suspended, unattached state
Prevents stem cells from attachment-mediated differentiation
Prevents anchorage-dependent cells from dividing
Reduces binding of attachment and serum proteins to the substrate
Chambers are sterilized by gamma radiation and certified nonpyrogenic. Manufactured under GMP conditions and 100% leak-tested prior to shipping.
The Corning® CellBIND® surface enhances cell attachment under difficult conditions, such as reduced-serum or serum-free medium, resulting in higher cell yields.
Accessories information: A variety of optional filling caps are available to allow direct aseptic transfer of media and cells via pumping or gravity feed. Several coupling devices are available on these filling caps with or without integrally sealed USP Class VI certified C-Flex® tubing. Optional filling caps with attached filters with hydrophobic membranes provide for gas exchange and faster aseptic venting during liquid transfers. Extra sterile vented or unvented 33 mm replacement caps are also available. Reusable stacking devices fit between CellSTACK® chambers to keep them level and optimise incubator space while providing clearance for gas exchange.
All caps are 33 mm threaded caps.
Documents
Specifications
- Recommended working volume:125 - 190 ml
- Seq:09
- Growth area:636 cm²
- Type:1 chamber
- Cat. No.:734-1017
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Universal cap, polyethylene, 33 mm, with vented overcap, double-bagged, sterileUniversal cap, polyethylene, 33 mm, with vented overcap, double-bagged, sterileCORN3732Log in for availability1 * 4 ST - 46,80 €
Product Information
- Catalog #AvailabilityPrice
- Solid cap, polyethylene, 33 mm, sterile
Product Information
- Catalog #AvailabilityPrice
- Vent cap, polyethylene, 33 mm, 0,2 µm membrane, sterileVent cap, polyethylene, 33 mm, 0,2 µm membrane, sterile734-4166Log in for availability1 * 6 ST - 51,30 €
Product Information
- Catalog #AvailabilityPrice
- Fill cap with male MPC; Int. Ø: 9,525 mm
Product Information
- Catalog #AvailabilityPrice
- Fill cap with male MPC; Int. Ø: 6,35 mm
Product Information
- Catalog #AvailabilityPrice
- Fill cap with female MPC coupling; Int. Ø: 9,525 mmFill cap with female MPC coupling; Int. Ø: 9,525 mm734-4063Log in for availability1 * 4 ST - 327,00 €
Product Information
- Catalog #AvailabilityPrice
- Fill cap with female MPC; Int. Ø: 6,35 mm
Product Information
- Catalog #AvailabilityPrice
- Two vented over caps and one solid over cap
Product Information
- Catalog #AvailabilityPrice
- Fill cap with male MPC; Int. Ø: 6,35 mm; Tubing length: 69,85 mmFill cap with male MPC; Int. Ø: 6,35 mm; Tubing length: 69,85 mm734-1209Log in for availability1 * 4 ST - 340,00 €
Product Information
- Catalog #AvailabilityPrice
- Vent cap with tubing and 37 mm hydrophobic glass laminate 1,0 μm vent filter; Int. Ø: 6,35 mm; Tubing length: 69,85 mmVent cap with tubing and 37 mm hydrophobic glass laminate 1,0 μm vent filter; Int. Ø: 6,35 mm; Tubing length: 69,85 mm734-1205Log in for availability1 * 4 ST - 442,00 €
Product Information
- Catalog #AvailabilityPrice
- Corning CellSTACK stacking device, ABS, nonsterileCorning CellSTACK stacking device, ABS, nonsterile734-4064Log in for availability1 * 5 ST - 246,00 €
Product Information
- Catalog #AvailabilityPrice
- Fill cap with tubing and barbed fittings; Int. Ø: 9,525 mm; Tubing length: 457,2 mmFill cap with tubing and barbed fittings; Int. Ø: 9,525 mm; Tubing length: 457,2 mm734-1110Log in for availability1 * 5 ST - 297,00 €
Product Information
- Catalog #AvailabilityPrice
- Fill cap with tubing, FLL; Int. Ø: 3,175 mm; Tubing length: 457,2 mmFill cap with tubing, FLL; Int. Ø: 3,175 mm; Tubing length: 457,2 mm734-1109Log in for availability1 * 5 ST - 256,00 €
Product Information
- Catalog #AvailabilityPrice
- Vent cap with tubing and 50 mm hydrophobic PVDF 0,2 µm vent filter; Int. Ø: 9,525 mm; Tubing length: 69,85 mmVent cap with tubing and 50 mm hydrophobic PVDF 0,2 µm vent filter; Int. Ø: 9,525 mm; Tubing length: 69,85 mm734-1108Log in for availability1 * 5 ST - 472,00 €
Product Information
- Catalog #AvailabilityPrice
- CellSTACK manifold - 4 Luer and 2 MPC connectionsCellSTACK manifold - 4 Luer and 2 MPC connectionsCORN11931Log in for availability1 * 4 ST - 589,00 €
Product Information
- Catalog #AvailabilityPrice
- Barbed cap with AseptiQuik® G series; Int. Ø: 9,525 mm; Tubing length: 609,6 mmBarbed cap with AseptiQuik® G series; Int. Ø: 9,525 mm; Tubing length: 609,6 mmCORN11759Log in for availability1 * 5 ST - 1 170,00 €
Product Information
- Catalog #AvailabilityPrice
- CellSTACK 2 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mmCellSTACK 2 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mmCORN11760Log in for availability1 * 8 ST - 2 290,00 €
Product Information
- Catalog #AvailabilityPrice
- CellSTACK 10 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mmCellSTACK 10 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mmCORN11762Log in for availability1 * 4 ST - 1 850,00 €
Product Information
- Catalog #AvailabilityPrice
- CellSTACK screw cap vent accessory with tubing and 50 mm hydrophobic PVDF 0,2 μm vent filter; Int. Ø: 9,525 mm; Tubing length: 76,2 mmCellSTACK screw cap vent accessory with tubing and 50 mm hydrophobic PVDF 0,2 μm vent filter; Int. Ø: 9,525 mm; Tubing length: 76,2 mm734-3409Log in for availability1 * 5 ST - 549,00 €
Product Information
- Catalog #AvailabilityPrice
- Barbed cap with female MPC; Int. Ø: 9,525 mm; Tubing length: 609,6 mmBarbed cap with female MPC; Int. Ø: 9,525 mm; Tubing length: 609,6 mmCORN11758Log in for availability1 * 5 ST - 634,00 €
Product Information
- Catalog #AvailabilityPrice
- CellSTACK 5 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mmCellSTACK 5 layer, female MPC; Tubing: chemically resistant, heat sealable flexible tubing; Int. Ø: 9,525 mm, Ext. Ø: 15,875 mm; Tubing length: 152,4 mmCORN11761Log in for availability1 * 4 ST - 1 530,00 €
Product Information
- Catalog #AvailabilityPrice
- CellSTACK screw cap fill accessory with tubing; Int. Ø: 9,525 mm; Tubing length: 381 mmCellSTACK screw cap fill accessory with tubing; Int. Ø: 9,525 mm; Tubing length: 381 mm734-3410Log in for availability1 * 5 ST - 360,00 €