ContactUsLinkComponent
CD 500 DIN,CADMIUM IONSELECTIVE ELECTRODE WITH 3.3 FT (1 M) CABLE, DIN PLUG, 0,01 TO 11,000 MG/L, REQUIRES REFERENCE R 503/D OR /P W ITH ELY/BR/503, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTIONS HAVE TO BE PREPARED FRESHLY. ACCESSORIES ARE NOT IN 1 * 1 items
Catalog # 662-0253
Supplier:  WTW
CD 500 DIN,CADMIUM IONSELECTIVE ELECTRODE WITH 3.3 FT (1 M) CABLE, DIN PLUG, 0,01 TO 11,000 MG/L, REQUIRES REFERENCE R 503/D OR /P W ITH ELY/BR/503, IONIC STRENGTH ADAPTER ISA/FK, STANDARD SOLUTIONS HAVE TO BE PREPARED FRESHLY. ACCESSORIES ARE NOT IN 1 * 1 items
Catalog # 662-0253
Supplier:  WTW
Frequently Bought Together