Buffer solutions, ready to use
10,0, Borate, Colourless, 20 °C, Plastic bottle, 5 L
Catalog # KRAF03089.5000
Supplier: Bernd Kraft
Jump to:
Product Details & Documents
- NIST traceable
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Plastic bottle
- pH buffer type:Borate
- Pk:5 L
- Reference temperature:20 °C
- Colour:Colourless
- pH value:10,0



