Master mix, BugBuster®
BugBuster® Master Mix, 500 mL
Catalog # 71456-4
Supplier: Merck Millipore (Novagen)
Jump to:
Product Details & Documents
BugBuster® master mix combines BugBuster® protein extraction reagent with Benzonase® Nuclease and rLysozyme™ solution in one convenient reagent. It allows for maximum recovery of active soluble protein from both Gram-negative and Gram-positive bacteria. The master mix provides sufficient reagents for protein extraction from 20 g and 100 g cell paste.
Specifications
Specifications
- Pk:500 mL
- Description:BugBuster® Master Mix
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



