Master mix, BugBuster®
BugBuster® Master Mix, 100 mL
Catalog # 71456-3
Supplier: Merck Millipore (Novagen)
Aller à :
Product Details & Documents
BugBuster® master mix combines BugBuster® protein extraction reagent with Benzonase® Nuclease and rLysozyme™ solution in one convenient reagent. It allows for maximum recovery of active soluble protein from both Gram-negative and Gram-positive bacteria. The master mix provides sufficient reagents for protein extraction from 20 g and 100 g cell paste.
Spécifications
Spécifications
- Pk:100 mL
- Description:BugBuster® Master Mix



