Order Entry
Belgium
Orders LinkContactUsLinkComponent
Petri dishes, Nunc™

Petri dishes, Nunc™

Petri dish, vented, 140 mm, 20 mm, Sterile
Catalog # 391-0103
   Low Manufacturing Impact
Petri dishes, Nunc™
Catalog # 391-0103
   Low Manufacturing Impact
Petri dishes, Nunc™
Petri dishes, Nunc™

Petri dish, vented, 140 mm, 20 mm, Sterile

Price
173,00 €
Log in for availability
1 * 80 ST
Product Details & Documents

Optically clear PS. Petri dishes for culturing fungi, bacteria and other microorganisms.

  • Compatible with automated systems
  • Perform well in automatic dispensers due to complete flatness and uniform height
Specifications
  • Ø:
    140 mm
  • Sterility:
    Sterile
  • Version:
    Petri dish, vented
  • Height:
    20 mm
  • Cat. No.:
    391-0103
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.