Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Flasks, round bottom, Quickfit® ground joints

100 ml, 24/29 NS, 110 mm
  201-8471
 :   SciLabware
Flasks, round bottom, Quickfit® ground joints
  201-8471
Flasks, round bottom, Quickfit® ground joints
Flasks, round bottom, Quickfit® ground joints

100 ml, 24/29 NS, 110 mm

 
110,00 €
   
1 * 5 ST
 

PYREX®, borosilicate glass, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With white, matt labelling area.

 : ISO 1773, ISO 4797.

 
  • Socket size:
    24/29 NS
  • Capacity:
    100 ml
  • Height:
    110 mm
  • Seq:
    14