Flasks, flat bottom with Quickfit® ground joint
250 ml, 125 mm, 29/32 NS
Catalog # 214-0048
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, clear, with short neck.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat base allows use with hotplates. Labelling area.
Specifications
- Socket size:29/32 NS
- Capacity:250 ml
- Height:125 mm
- Seq:13



