Counting chambers, Neubauer
Counting chamber, dark-lined, with clamps, 0,10 mm
Supplier: MARIENFELD
Jump to:
Product Details & Documents
These counting chambers are designed for counting leucocytes and erythrocytes.
- Double net ruling
- Two counting grids with one division
Certifications: IVD 98/79/EC
Delivery information: Supplied with two cover glasses.
Documents
Certificates
Search by Lot NumberSpecifications
- Depth of chamber:0,10 mm
- Description:Counting chamber, dark-lined, with clamps
- Seq:04