Order Entry
Belgium
Orders LinkContactUsLinkComponent
Weighing scoops

Weighing scoops

10 ml, 116 mm
Catalog # 611-9322
Supplier:   Lenz Laborglas
Weighing scoops
Catalog # 611-9322
Supplier:  Lenz Laborglas
Weighing scoops
Weighing scoops

10 ml, 116 mm

Price
26,80 €
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass.

  • Ideal for weighing small samples
  • The tubular stem of the scoop also works as a weighing funnel and facilitates emptying of the scoop

Documents

Specifications
  • Capacity:
    10 ml
  • Length:
    116 mm