Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pestles

45 mm, 180 mm, 410-9327
  410-0077
Pestles
  410-0077
Pestles
Pestles

45 mm, 180 mm, 410-9327

 
72,60 €
   
1 * 1 ST
 

Porcelain with glazed handle, glazed or unglazed grinding surface.

 : Manufactured to DIN 12906.

 
  • For:
    410-9327
  • Head Ø:
    45 mm
  • Length:
    180 mm
  • Seq:
    06