Order Entry
Belgium
Orders LinkContactUsLinkComponent
Pestles

Pestles

30 mm, 135 mm, 410-9328
Catalog # 410-0075
Pestles
Catalog # 410-0075
Pestles
Pestles

30 mm, 135 mm, 410-9328

Price
44,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Porcelain with glazed handle, glazed or unglazed grinding surface.

Certifications: Manufactured to DIN 12906.

Specifications
  • For:
    410-9328
  • Head Ø:
    30 mm
  • Length:
    135 mm
  • Seq:
    03