Orders LinkContactUsLinkComponent
Volumetric Flasks, Class A
2000 ml, 19/26, 0,60 ± ml
Catalog # WITG3.664.200.BL
Supplier:   witeg Labortechnik
Volumetric Flasks, Class A
Catalog # WITG3.664.200.BL
Supplier:  witeg Labortechnik
Volumetric Flasks, Class A
Volumetric Flasks, Class A
2000 ml, 19/26, 0,60 ± ml
Price
116,00 €
Log in for availability
1 * 2 ST
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
  • DE-M marked according to the German calibration law
  • Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifications
  • NS:
    19/26
  • Capacity:
    2000 ml
  • Seq:
    15
  • Tolerance:
    0,60 ± ml