Orders LinkContactUsLinkComponent
Adapters, reduction, Preciso
Adapters, reduction, Preciso
Reduction, 29/32 NS, 19/26
Catalog # WITG0.500.005
Supplier:   witeg Labortechnik
Adapters, reduction, Preciso
Catalog # WITG0.500.005
Supplier:  witeg Labortechnik
Adapters, reduction, Preciso
Adapters, reduction, Preciso
Reduction, 29/32 NS, 19/26
Price
9,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
Certifications: According to DIN 12257.
Specifications
  • Cone size:
    29/32 NS
  • Socket:
    19/26
  • Type:
    Reduction