Orders LinkContactUsLinkComponent
Certified colour reference standards
Certified colour reference standards
AOCS-Tintometer® Colour set for colorimeter AF710-3, 1
Catalog # 633-0258
Supplier:   Lovibond Tintometer
Certified colour reference standards
Catalog # 633-0258
CAS Number:  
Certified colour reference standards
Certified colour reference standards
AOCS-Tintometer® Colour set for colorimeter AF710-3, 1
CAS Number:  
Price
2 020,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
For routine calibration of colorimeters and verification of test data.
Lovibond® certified colour reference materials are supplied with full traceability to internationally recognised standards. Each standard comes in a 500 ml bottle with a 12-month guarantee of colour stability and full certification including MSDS. Individual values may vary but these are always specified on the certificate of calibration supplied with each standard.
Documents
Specifications
Specifications
  • Pk:
    1
  • Description:
    AOCS-Tintometer® Colour set for colorimeter AF710-3