Lovibond RYBN 1.0R 4.3Y 0.1N (5 1/4) Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0049
Supplier: Lovibond Tintometer
Lovibond RYBN 1.0R 4.3Y 0.1N (5 1/4) Certified Colour Reference Standard 1 * 500 mL
Catalog # 633-0049
Supplier: Lovibond Tintometer
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Water Reag. Ph. Eur. 1095506 nitrate-free
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
VWR®, General purpose viscosity standards
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
Turbidity standards, Stablcal®
Supplier: Hach
n-Tetradecane, Supelco®
Supplier: Merck