Orders LinkContactUsLinkComponent
Tripod stands
Tripod stands
120×250 mm
Catalog # 451-0157
Supplier:   USBECK Laborgeräte
Tripod stands
Catalog # 451-0157
Tripod stands
Tripod stands
120×250 mm
Price
22,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.
Specifications
  • Int.Ø×H:
    120×250 mm
  • Seq:
    09