Orders LinkContactUsLinkComponent
Dissection sets
Dissection sets
Small microscopy set, 6 pcs.
Catalog # 631-0616
Supplier:   RSG
Dissection sets
Catalog # 631-0616
Supplier:  RSG
Dissection sets
Dissection sets
Small microscopy set, 6 pcs.
Price
38,70 €
Log in for availability
1 * 1 SET
Product Details & Documents
Stainless steel. These instruments are designed for the dissection of samples ready for microscopic analysis.
Small set - 6 pcs. Scalpel pointed, 160 mm, blade 40 mm; spatula, 150×10 mm; tweezer pointed, straight, 105 mm; microscopy scissors, straight, 100 mm; needle straight with plastic handle, 140 mm; lance with plastic handle, 140 mm
Large set - 9 pcs. Scalpel pointed, 160 mm, blade 40 mm; spatula 150×10 mm; tweezer pointed, straight, 105 mm; microscopy scissors, straight, 100 mm; microscopy scissors, curved, 100 mm; needle straight with plastic handle, 140 mm; needle curved with plastic handle, 140 mm; lance with plastic handle, 140 mm; razor with blade and plastic handle
Anatomical set - 13 pcs. Scalpel pointed, 160 mm, blade 40 mm; scalpel pointed, 130 mm, blade 17 mm; scalpel round, 130 mm, blade 17 mm; spatula 150×10 mm; tweezer pointed, straight, 105 mm; tweezer blunt, straight, 115 mm; microscopy scissors, straight, 100 mm; microscopy scissors, curved, 100 mm; bandage scissors pointed/blunt, 130 mm; director, 150 mm; needle straight with plastic handle, 140 mm; needle curved with plastic handle, 140 mm; lance with plastic handle, 140 mm
Delivery information: Supplied in leather case with zipper.
Specifications
  • Description:
    Small microscopy set, 6 pcs.