Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

150 mm, 2 mm
Catalog # 231-0461
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0461
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro

150 mm, 2 mm

Price
17,40 €
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.

  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades

Documents

Specifications
  • Length:
    150 mm
  • Blade width:
    2 mm