Order Entry
Belgium
Orders LinkContactUsLinkComponent
Double spatulas, micro

Double spatulas, micro

100 mm, 5 mm
Catalog # 231-0466
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0466
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro

100 mm, 5 mm

Price
31,70 €
Log in for availability
1 * 5 ST
Product Details & Documents

Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.

  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades

Documents

Specifications
  • Length:
    100 mm
  • Blade width:
    5 mm