Order Entry
Belgium
Orders LinkContactUsLinkComponent
Support rods, polished

Support rods, polished

12 mm, 1500 mm
Catalog # RSGA241.210
Supplier:   RSG
Support rods, polished
Catalog # RSGA241.210
Supplier:  RSG
Support rods, polished
Support rods, polished

12 mm, 1500 mm

Price
79,30 €
Log in for availability
1 * 1 ST
Product Details & Documents

Polished rods made from stainless steel 18/10. Available with or without thread.

Specifications
  • Ø:
    12 mm
  • Length:
    1500 mm
  • Seq:
    14
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.